missing translation for 'onlineSavingsMsg'
Learn More

Neprilysin/CD10 Antibody (MME/8376R), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 286.00 - € 530.00

Specifications

Antigen Neprilysin/CD10
Clone MME/8376R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18660394
View Documents
Novus Biologicals
NBP3-20635-100ug
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18690215
View Documents
Novus Biologicals
NBP3-20635-20ug
20 μg
€ 286.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Neprilysin/CD10 Monoclonal antibody specifically detects Neprilysin/CD10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Neprilysin/CD10
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers
10mM PBS with 0.05% BSA
4311
IgG κ
Protein A or G purified
MME/8376R
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
Atriopeptidase, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11, SFE, Skin fibroblast elastase
Recombinant human Neprilysin/CD10 protein fragment (aa 1-500) (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.