missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neprilysin/CD10 Antibody (OTI3D11), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72941R
This item is not returnable.
View return policy
Description
Neprilysin/CD10 Monoclonal specifically detects Neprilysin/CD10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Neprilysin/CD10 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MME | |
| Full length recombinant protein of human Neprilysin/CD10 (NP_009218) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI3D11 | |
| Western Blot, Immunohistochemistry | |
| Atriopeptidase, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11, SFE, Skin fibroblast elastase | |
| Mouse | |
| Affinity Purified | |
| B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
| 4311.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction