missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester, 95%

Merk:  Alfa Aesar™ H63470.03

Overige gegevens : CAS : 34404-36-9 Gewicht : 0.00100kg

Aantal 1g

 Bekijk meerdere types van dit product via de

Produktcode. 15448326

  • € 22.40 / 1 gram


Geschatte verzenddatum
bij leverancier 15-04-2019
Toevoegen aan winkelmandje

Beschrijving

Beschrijving

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specificatie

Chemische identificatiekenmerken

34404-36-9
477.514
YWLICOCXPNQJPC-KRWDZBQOSA-N
12686578
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
C23H31N3O8
MFCD00037915
boc-lys z-osu, a-boc-l-lysine hydroxysuccinimide ester, n,a-boc-n, a-cbz-l-lysine n-hydroxysuccinimide ester, s-2,5-dioxopyrrolidin-1-yl 6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys cbz-osu, 2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys z osu, boc-lys z-o su, boc-lys-z-osu, n,a-boc-n, a-z-l-lysine hydroxysuccinimide ester, n,a-z-n
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoate

Specificatie

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester
C23H31N3O8
1559393
YWLICOCXPNQJPC-KRWDZBQOSA-N
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoate
12686578
Powder
White
1g
34404-36-9
MFCD00037915
boc-lys z-osu, a-boc-l-lysine hydroxysuccinimide ester, n,a-boc-n, a-cbz-l-lysine n-hydroxysuccinimide ester, s-2,5-dioxopyrrolidin-1-yl 6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys cbz-osu, 2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys z osu, boc-lys z-o su, boc-lys-z-osu, n,a-boc-n, a-z-l-lysine hydroxysuccinimide ester, n,a-z-n
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
477.514
477.51
95%
108°C to 117°C
Video's
SDS