missing translation for 'onlineSavingsMsg'
Learn More

Neqoinate, TRC

Product Code. 30138162
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138162 25 mg 25mg
30006382 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138162 Supplier Toronto Research Chemicals Supplier No. TRCN38993025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13997-19-8
Molecular Formula C22 H23 N O4
Molecular Weight (g/mol) 365.42
Synonym 3-Quinolinecarboxylic acid, 6-butyl-1,4-dihydro-4-oxo-7-(phenylmethoxy)-, methyl ester3-Quinolinecarboxylic acid, 7-(benzyloxy)-6-butyl-1,4-dihydro-4-oxo-, methyl ester (8CI), 7-Benzyloxy-6-butyl-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid methyl ester, AY 20385, ICI 55052, Mequinate, Methyl 7-(benzyloxy)-6-butyl-1,4-dihydro-4-oxo-3-quinoline carboxylate, Methyl 7-(benzyloxy)-6-n-butyl-4-oxo-1,4-dihydroquinoline-3-carboxylate, Methyl benzoquate, Neqoinate, Nequinate, Statil, Statoquate, Statyl, Tranil
IUPAC Name methyl 6-butyl-4-oxo-7-phenylmethoxy-1H-quinoline-3-carboxylate
SMILES CCCCc1cc2C(=O)C(=CNc2cc1OCc3ccccc3)C(=O)OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H23NO4/c1-3-4-10-16-11-17-19(23-13-18(21(17)24)22(25)26-2)12-20(16)27-14-15-8-6-5-7-9-15/h5-9,11-13H,3-4,10,14H2,1-2H3,(H,23,24)
Formula Weight 365.1627
Chemical Name or Material Nequinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.