missing translation for 'onlineSavingsMsg'
Learn More

Neratinib, MedChemExpress

Product Code. 16403967
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16403967

Brand: Medchem Express HY32721

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neratinib (HKI-272) is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively.

Neratinib (HKI-272) is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively.

Specifications

Chemical Name or Material Neratinib
CAS 698387-09-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H29ClN6O3
Quantity 50 mg
Synonym HKI-272
Solubility Information DMSO : 13.33 mg/mL (23.93 mM; Need ultrasonic)
SMILES ClC1=C(OCC2=NC=CC=C2)C=CC(NC3=C(C#N)C=NC4=CC(OCC)=C(NC(/C=C/CN(C)C)=O)C=C43)=C1
Molecular Weight (g/mol) 557.04
Formula Weight 557.04
Percent Purity 96.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.