Learn More
Nerolidol (Mixture of cis and trans, Stablized with Alpha-tocopherol), TRC
Product Code. 30466976
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466976 5 g -
30471604 100 g 100g
30469312 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30466976 Supplier Toronto Research Chemicals Supplier No. TRCN3901205G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7212-44-4
Molecular Formula C15 H26 O
Molecular Weight (g/mol) 222.37
Synonym 1,6,10-Dodecatrien-3-ol, 3,7,11-trimethyl-, Nerolidol (6CI), 3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, (±)-Nerolidol, FCI 119b, Nerodilol
IUPAC Name 3,7,11-trimethyldodeca-1,6,10-trien-3-ol
SMILES CC(=CCCC(=CCCC(C)(O)C=C)C)C
Specifications
Quantity 5 g
InChI Formula InChI=1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3
Formula Weight 222.1984
Chemical Name or Material (cis + trans)-Nerolidol