missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nesfatin-1/Nucleobindin-2 Antibody (2062A), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB59491G
This item is not returnable.
View return policy
Description
Nesfatin-1/Nucleobindin-2 Monoclonal specifically detects Nesfatin-1/Nucleobindin-2 in Human samples. It is validated for Western Blot.
Specifications
| Nesfatin-1/Nucleobindin-2 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NUCB2 | |
| Partial recombinant human NUCB2/Nesfatin-1 protein (amino acids 25-106) [UniProt P80303] | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 2062A | |
| Western Blot | |
| DNA-binding protein NEFA, NEFAGastric cancer antigen Zg4, nucleobindin 2, nucleobindin-2, nucleobinding 2 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 4925 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction