missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nesprin-3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33295-100ul
This item is not returnable.
View return policy
Description
Nesprin-3 Monoclonal antibody specifically detects Nesprin-3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Nesprin-3 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| chromosome 14 open reading frame 49, FLJ25605, MGC75267, nesprin-3, NET53 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 750-935 of human Nesprin-3 (NP_689805.3).,, Sequence:, LIRNWHLQRMEVDSGKKMVFTNNIPKSGFLINPMDPIPRHRRRANLLQEEEGSHEDFSQLLRNFGQWLQVENSKLVRIIAMRTSTAEDLRTRKSKLQELEARVPEGQHLFENLLRLGPARGTSDELEDLRYQWMLYKSKLKDSGHLLTQSSPGEPTGFQKTRRWRGLGSLFRRACCVALPLQLLLL | |
| 100 μL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 161176 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction