missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nesprin-3 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33295-20ul
This item is not returnable.
View return policy
Description
Nesprin-3 Monoclonal antibody specifically detects Nesprin-3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Nesprin-3 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| chromosome 14 open reading frame 49, FLJ25605, MGC75267, nesprin-3, NET53 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 750-935 of human Nesprin-3 (NP_689805.3).,, Sequence:, LIRNWHLQRMEVDSGKKMVFTNNIPKSGFLINPMDPIPRHRRRANLLQEEEGSHEDFSQLLRNFGQWLQVENSKLVRIIAMRTSTAEDLRTRKSKLQELEARVPEGQHLFENLLRLGPARGTSDELEDLRYQWMLYKSKLKDSGHLLTQSSPGEPTGFQKTRRWRGLGSLFRRACCVALPLQLLLL | |
| 20 μL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 161176 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?