Learn More
Nesprin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27132-1-AP-150UL
This item is not returnable.
View return policy
Description
Component of SUN-protein-containing multivariate complexes also called LINC complexes which link the nucleoskeleton and cytoskeleton by providing versatile outer nuclear membrane attachment sites for cytoskeletal filaments. Involved in the maintece of nuclear organization and structural integrity. Probable anchoring protein which tethers the nucleus to the cytoskeleton by binding PLEC which can associate with the intermediate filament system.Specifications
| Nesprin 3 | |
| Polyclonal | |
| Unconjugated | |
| SYNE3 | |
| C14orf49, Nesprin 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 161176, 212073, 299356 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.77 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4FZC9, Q6ZMZ3 | |
| Syne3 | |
| Nesprin 3 Fusion Protein Ag26002 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |