missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nestin Antibody (10C2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-266C
This item is not returnable.
View return policy
Description
Nestin Monoclonal specifically detects Nestin in Human, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Nestin | |
| Monoclonal | |
| DyLight 650 | |
| P48681 | |
| NES | |
| A fragment of the human Nestin protein corresponding to residues 1464-1614. [UniProt P48681] | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 10763 | |
| Human, Mouse (Negative), Rabbit (Negative), Rat (Negative) | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 10C2 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| FLJ21841, nestin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human. It does not cross-react mouse, rat or rabbit. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction