missing translation for 'onlineSavingsMsg'
Learn More

Nestin Antibody, Novus Biologicals™

Chicken Polyclonal Antibody has been used in 46 publications

Brand:  Novus Biologicals NB100-1604-0.05ml

 View more versions of this product

Product Code. 18572504

  • ONLINE EXCLUSIVE PRICE
    € 421.00
    € 397.95 / 50µL
    Save € 23.05
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nestin Polyclonal specifically detects Nestin in Human, Mouse, Rat, Baboon, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nestin
Polyclonal
Unconjugated
10 mM Sodium PBS (0.9% isotonic, w/v, pH 7.2) with 0.02% Sodium Azide
FLJ21841, nestin
Chicken
Affinity Purified
RUO
Primary
Nestin
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
0.1 mg/mL
Western Blot 1:10000 - 1:20000, Flow Cytometry, Immunohistochemistry 1:1000 - 1:2000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunohistochemistry-Paraffin 1:1000 - 1:2000, Immunohistochemistry-Frozen 1:1000 - 1:2000
Q6P5H2, NP_057910
NES
Chickens were immunized with three synthetic peptide/keyhole limpet hemocyanin (KLH) conjugates. These synthetic peptides corresponded to different regions of the mouse nestin gene product (NM_NP_057910), but are shared with the rat (NM_012987, NCBI) protein sequence.
0.05 mL
Stem Cell Markers
10763
Human, Mouse, Rat, Baboon, Monkey
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only