missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nestin Monoclonal Antibody (3k1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA126777

Product Code. 11539552

  • € 600.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: Human neural progenitors and A172 cells. MA1-26777 will not react with rodent. Keep as concentrated solution. Store at 4°C short term. For extended storage aliquot- add 50% glycerol and store at -20°C or below. Avoid freeze-thaw cycles.

Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nestin
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative; pH 7.4
P48681, Q6P5H2
Nes
NS0 cells transfected with human Nestin.
50 μg
Primary
Human, Mouse
Antibody
IgG1
Flow Cytometry, Western Blot, Immunocytochemistry
3k1
Unconjugated
Nes
AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin
Mouse
Protein G
RUO
10763, 18008
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.