missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nestin Monoclonal Antibody (4D11)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547329
This item is not returnable.
View return policy
Description
Nestin Monoclonal Antibody for Western Blot, ICC/IF, IHC
Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.
Specifications
| Nestin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P21263, P48681, Q6P5H2 | |
| Nes | |
| Recombinant construct, amino acids 317-630 of the human protein expressed in and purified from E. coli. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 4D11 | |
| Unconjugated | |
| Nes | |
| AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin | |
| Mouse | |
| Protein G | |
| RUO | |
| 10763, 18008, 25491 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction