missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nestin Polyclonal Antibody
GREENER_CHOICE

Chicken Polyclonal Antibody

€ 504.00 - € 1736.00

Specifications

Antigen Nestin
Concentration Conc. Not Determined
Content And Storage 4°C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17866777
View Documents
Invitrogen™
PA5143544
500 μL
€ 1736.00
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17886797
GREENER_CHOICE
View Documents
Invitrogen™
MA547459
100 μL
€ 504.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nestin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.
Specifications

Specifications

Nestin
4°C
Polyclonal
Liquid
IgY
Human, Mouse, Rat
Nes
AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin
Nes
Primary
Conc. Not Determined
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Chicken
RUO
PBS with 0.02% sodium azide
P21263, P48681, Q6P5H2
10763, 18008, 25491
Recombinant construct, amino acids 317-630 of the human protein expressed in and purified from E. coli.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.