missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NETO1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41134H
This item is not returnable.
View return policy
Description
NETO1 Polyclonal antibody specifically detects NETO1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NETO1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1, BTCL1BCTL1neuropilin and tolloid-like protein 1, FLJ41325, neuropilin (NRP) and tolloid (TLL)-like 1 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human NETO1. The immunogen is located within amino acids 350 - 400 of NETO1. Amino Acid Squence: KQPRKKYVQRKSDFDQTV | |
| 0.1 mL | |
| Vision | |
| 81832 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction