missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NETO1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | NETO1 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639376
|
Novus Biologicals
NBP2-99822-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635126
|
Novus Biologicals
NBP2-99822-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NETO1 Polyclonal antibody specifically detects NETO1 in Human samples. It is validated for ELISASpecifications
| NETO1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| 0.2 um filtered solution in PBS | |
| 81832 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 1, BTCL1BCTL1neuropilin and tolloid-like protein 1, FLJ41325, neuropilin (NRP) and tolloid (TLL)-like 1 | |
| Produced in rabbits immunized with purified, recombinant Human NETO1 (Accession#: NP_620416.1; Met 1-Thr 344) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title