missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Netrin G1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12910-2-AP-20UL
This item is not returnable.
View return policy
Description
Netrin G1.Specifications
| Netrin G1 | |
| Polyclonal | |
| Unconjugated | |
| NTNG1 | |
| KIAA0976, Laminet 1, Lmnt1, Netrin G1, NTNG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22854, 295382, 80883 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8R4G0, Q9Y2I2 | |
| Ntng1 | |
| Netrin G1 Fusion Protein Ag3882 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction