missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Netrin-G1a Antibody (1010035), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10019CL1
This item is not returnable.
View return policy
Description
Netrin-G1a Monoclonal antibody specifically detects Netrin-G1a in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Netrin-G1a | |
| Monoclonal | |
| CoraFluor 1 | |
| Lmnt1, NTNG1 | |
| Human embryonic kidney cell, HEK293-derived human Netrin-G1a, His29-Gly503, Accession # Q9Y212 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 1010035 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 22854 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction