missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEU1 Monoclonal Antibody (OTI3B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
POA
Specifications
| Antigen | NEU1 |
|---|---|
| Clone | OTI3B8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17570063
|
Origene Technologies
TA801671S |
30 μL |
POA
30µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17550063
|
Origene Technologies
TA801671 |
100 μL |
POA
100µL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NEU1 Monoclonal antibody specifically detects NEU1 in Human samples. It is validated for Western BlotSpecifications
| NEU1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q99519 | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
| OTI3B8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 4758 | |
| Primary | |
| NEU1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title