missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEU3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545518
This item is not returnable.
View return policy
Description
Peptide sequence: EEEGLFGCLF ECGTKQECEQ IAFRLFTHRE ILSHLQGDCT SPGRNPSQFK Sequence homology: Horse: 100%; Human: 100%.
The function of this protein remains unknown.
Specifications
| NEU3 | |
| Polyclonal | |
| Unconjugated | |
| NEU3 | |
| Ganglioside sialidase; LOW QUALITY PROTEIN: sialidase-3; membrane sialidase; N-acetyl-alpha-neuraminidase 3; NEU3; neuraminidase 3; neuraminidase 3 (membrane sialidase); neuraminidase 3, membrane sialidase; SIAL3; sialidase 3 (membrane sialidase); sialidase-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10825 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UQ49 | |
| NEU3 | |
| Synthetic peptide directed towards the C-terminal of human NEU3 (aa 377-426). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction