missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEU4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12995-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to a family of glycohydrolytic enzymes, which remove terminal sialic acid residues from various sialo derivatives, such as glycoproteins, glycolipids, oligosaccharides, and gangliosides. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Specifications
| NEU4 | |
| Polyclonal | |
| Unconjugated | |
| NEU4 | |
| N acetyl alpha neuraminidase 4, NEU4, sialidase 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 129807 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WWR8 | |
| NEU4 | |
| NEU4 Fusion Protein Ag3657 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction