missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NeuN Monoclonal Antibody (K06325_3H12), Invitrogen™
Mouse Monoclonal Antibody
€ 524.00 - € 876.00
Specifications
| Antigen | NeuN |
|---|---|
| Clone | K06325_3H12 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30196514
|
Invitrogen™
MA556047 |
200 μL |
€ 876.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30209867
|
Invitrogen™
MA555667 |
50 μL |
€ 524.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: AQTHPEQPGS EASTQPIAGT QTVPQTDEAA QTDSQPLHPS DPTEKQQPKR LHVSNIPFRF RDPDLRQMFG QFGKILDVEI IFNERGSKGF GFVTFETSSD ADRAREKLNG TIVEGRKIEV NNATARVMTN KKTGNPYTNG
NeuN Monoclonal antibody specifically detects NeuN in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| NeuN | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3 | |
| Recombinant human RBFOX3 (immunogen range is 51-190). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| RBFOX3 |
| K06325_3H12 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| A6NFN3 | |
| 146713, 287847 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title