missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537407
Additional Details : Weight : 0.00010kg
Descripción
This antibody detects endogenous protein at a molecular weight of 38 kDa. Purity is >95% by SDS-PAGE.
NeuN Polyclonal antibody specifically detects NeuN in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
NeuN | |
Polyclonal | |
Unconjugated | |
RBFOX3 | |
D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146713, 287847, 52897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
A6NFN3, Q8BIF2 | |
RBFOX3 | |
Recombinant full length human NeuN | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |