Learn More
Invitrogen™ NeuN Recombinant Rabbit Monoclonal Antibody (RM312)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA544596
Product Code. 17896771
  • € 452.00 / 100µL
Description
NeuN Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Fox-3, also known as NEUN, HRNBP3, RBFOX3 (RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NeuN
Recombinant Monoclonal
100 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
A6NFN3
RBFOX3
A peptide corresponding to the N-terminus of human NeuN (Fox3, RBFOX3).
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
RM312
Unconjugated
RBFOX3
D11Bwg0517e; FLJ56884; FLJ58356; fox 1 homolog (C. elegans) 3; fox1 homolog C; Fox-1 homolog C; FOX3; FOX-3; FOX3NeuN; hexaribonucleotide binding protein 3; hexaribonucleotide binding protein 3 (HRNBP3); Hexaribonucleotide-binding protein 3; Hrnbp3; NEUN; NeuN antigen; Neuna60; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RBFOX3; RFOX3; RGD1560070; RNA binding fox-1 homolog 3; RNA binding protein; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3
Rabbit
Protein A
RUO
146713
-20°C