missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuregulin-1/NRG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19588
This item is not returnable.
View return policy
Description
Neuregulin-1/NRG1 Polyclonal specifically detects Neuregulin-1/NRG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Neuregulin-1/NRG1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF | |
| Rabbit | |
| 70 kDa | |
| 0.1 mL | |
| Biologically Active Proteins, Chromatin Research, Glia Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction | |
| 3084 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q02297 | |
| NRG1 | |
| Recombinant protein encompassing a sequence within the center region of human Neuregulin-1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction