missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neurexin-1-beta Polyclonal Antibody
GREENER_CHOICE

Product Code. 30534916
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534916 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534916 Supplier Invitrogen™ Supplier No. PA5148861

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Neurexin-1-beta (NRXN1-beta) is a gene that encodes the beta-isoform of the neurexin-1 protein, which is a type I membrane protein that functions as a cell adhesion molecule and receptor in the nervous system. Located on chromosome 2 in humans, the neurexin-1 gene undergoes complex alternative splicing, producing multiple isoforms, including the well-studied alpha-neurexin and beta-neurexin. Neurexins, including NRXN1-beta, are primarily involved in synapse formation and function by mediating interactions between presynaptic and postsynaptic neurons. They bind to neuroligins, another family of cell adhesion molecules, thus playing a critical role in the maintenance and regulation of synaptic specificity and neurotransmitter release. Dysregulation or mutations in NRXN1-beta have been implicated in various neurological and psychiatric disorders, including autism spectrum disorders, schizophrenia, and intellectual disabilities, emphasizing the importance of this gene in brain development and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurexin-1-beta
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene NRXN1
Gene Accession No. Q63372
Gene Alias 1700062G21Rik; 9330127H16Rik; A230068P09Rik; alpha-latrotoxin receptor (calcium-dependent); DKFZp313P2036; FLJ35941; Hs.22998; Kiaa0578; mKIAA0578; neurexin 1; Neurexin 1alpha; neurexin I; neurexin I-alpha; Neurexin I-beta; neurexin-1; Neurexin-1-alpha; neurexin-1-beta; non-processed neurexin I-alpha; Nrxn1; Nrxn1b; PTHSL2; SCZD17
Gene Symbols NRXN1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neurexin 1B/NXRN1b, Ala47-Thr392.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60391
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.