missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEURL2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15958994
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15958994 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15958994 Supplier Invitrogen™ Supplier No. PA566283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YELNVLPPTAR RSRLGVLFCP RPDGTADMHI IINGEDMGPS ARGLPAAQPL YAVVDVFAST KSVRLVQLEY GL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

This gene encodes a protein that is involved in the regulation of myofibril organization. This protein is likely the adaptor component of the E3 ubiquitin ligase complex in striated muscle, and it regulates the ubiquitin-mediated degradation of beta-catenin during myogenesis. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEURL2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEURL2
Gene Accession No. Q9BR09
Gene Alias C20orf163; Neur2; neuralized E3 ubiquitin protein ligase 2; neuralized homolog 2; neuralized-like 2; neuralized-like 2 (Drosophila); neuralized-like protein 2; NEURL2; Ozz; Ozz-E3
Gene Symbols NEURL2
Host Species Rabbit
Immunogen Recombinant Human NEURL2. Recombinant protein control fragment (Product #RP-106968).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140825
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.