missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurobeachin Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | Neurobeachin |
|---|---|
| Concentration | 230 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235880
|
Proteintech
55287-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236097
|
Proteintech
55287-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Neurobeachin antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Neurobeachin. This antibody recognizes human, mouse antigen. The Neurobeachin antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Neurobeachin | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 230 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 26960 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title