missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuroD2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26068
This item is not returnable.
View return policy
Description
NeuroD2 Polyclonal specifically detects NeuroD2 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| NeuroD2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| NEUROD2 | |
| Peptide with sequence C-DGEDDEPRSDKGDAP corresponding to internal region (near N-Terminus) according to NP_006151.3. | |
| 0.1 mg | |
| Neuroscience | |
| 4761 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:100 - 1:2000, Peptide ELISA Detection limit 1:2000 | |
| BHLHA1, bHLHa1neurogenic differentiation factor 2, Class A basic helix-loop-helix protein 1, MGC26304, NDRFneurogenic basic-helix-loop-helix protein, NeuroD2, NeuroD-related factor, neurogenic differentiation 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction