missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEUROD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568245
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Probably acts as a transcriptional activator. Mediates neuronal differentiation. Required for the regulation of amacrine cell fate specification in the retina.
Specifications
| NEUROD4 | |
| Polyclonal | |
| Unconjugated | |
| NEUROD4 | |
| AI846749; ath3; ATH-3; Atoh3; atonal homolog 3; atonal-like 3; atonal-like protein 3; bHLH domain; bHLHa4; Class A basic helix-loop-helix protein 4; helix-loop-helix protein mATH-3; mATH3; MATH-3; NeuroD4; neurod4 protein; neurogenic differentiation 4; neurogenic differentiation factor 4; neuronal basic helix-loop-helix transcription factor Ath3; neuronal differentiation 4; protein atonal homolog 3; zath3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11923, 288821, 58158 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O09105, Q9HD90 | |
| NEUROD4 | |
| Synthesized peptide derived from human NEUROD4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction