missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofascin Antibody (1018843), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8208PECY7
This item is not returnable.
View return policy
Description
Neurofascin Monoclonal antibody specifically detects Neurofascin in Human samples. It is validated for Flow Cytometry
Specifications
| Neurofascin | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1018843 | |
| Flow Cytometry | |
| DKFZp686P2250, KIAA0756FLJ46866, neurofascin, neurofascin homolog, neurofascin homolog (chicken), NF, NRCAML | |
| Human embryonic kidney cell, HEK293-derived human Neurofascin, Ile25-Trp1039, Accession # NP_055905 | |
| 0.1 mL | |
| Neuroscience | |
| 23114 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction