missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neurofascin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102681
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NFASC.
Neurofascin encodes an L1 family immunoglobulin cell adhesion molecule with multiple IGcam and fibronectin domains. The protein functions in neurite outgrowth, neurite fasciculation, and organization of the axon initial segment (AIS) and nodes of Ranvier on axons during early development. Both the AIS and nodes of Ranvier contain high densities of voltage-gated Na+ (Nav) channels which are clustered by interactions with cytoskeletal and scaffolding proteins including this protein, gliomedin, ankyrin 3 (ankyrin-G), and betaIV spectrin. This protein links the AIS extracellular matrix to the intracellular cytoskeleton. Neurofascin undergoes extensive alternative splicing, and the full-length nature of some variants has not been determined.
Specifications
| Neurofascin | |
| Polyclonal | |
| Unconjugated | |
| NFASC | |
| AA387016; D430023G06Rik; DKFZp686P2250; KIAA0756; KIAA0756FLJ46866; mKIAA0756; neurofascin; neurofascin homolog; NF; Nfasc; NRCAML | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116690, 23114, 269116 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94856, P97685, Q810U3 | |
| NFASC | |
| A synthesized peptide derived from human NFASC(Accession O94856), corresponding to amino acid residues G1313-A1347. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction