missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofilament (NEFL) Monoclonal Antibody (OTI2C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF814553
This item is not returnable.
View return policy
Description
Neurofilament (NEFL) Monoclonal antibody specifically detects Neurofilament (NEFL) in Human samples. It is validated for ELISASpecifications
| Neurofilament (NEFL) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P14136 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2670 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| ELISA | |
| OTI2C12 | |
| Unconjugated | |
| GFAP | |
| 68 kDa neurofilament protein; AI847934; CMT1F; CMT2E; FLJ53642; light molecular weight neurofilament protein; micro glutamic acid-rich protein; NEF3; Nefl; nefl.L; neurofilament; neurofilament 3; neurofilament L subunit; neurofilament light; neurofilament light polypeptide; neurofilament protein; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; neurofilament triplet L protein; neurofilament, light L homeolog; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament, light polypeptide L homeolog; neurofilament-l; neurofilament-light; Nf68; NFL; NF-L; NFM; NF-M; NLC; OTTHUMP00000196614; PPP1R110; protein phosphatase 1, regulatory subunit 110; XELAEV_18018673mg; XNF-L | |
| Full length human recombinant protein of human GFAP (NP_002046) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction