missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEUROG1 Mouse anti-Human, Clone: OTI6C9, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500306
This item is not returnable.
View return policy
Description
Neurogenin 1 is a basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.Specifications
| NEUROG1 | |
| Monoclonal | |
| 0.75 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92886 | |
| NEUROG1 | |
| Full length human recombit protein of human NEUROG1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| OTI6C9 | |
| Unconjugated | |
| NEUROG1 | |
| AKA, Math4C, NEUROD3, bHLHa6, ngn1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4762 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction