Learn More
Invitrogen™ Neurogenin 1 Monoclonal Antibody (OTI3F9)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA524900
Product Code. 15984504
  • € 592.00 / 100µL
Description
Neurogenin 1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Neurogenin 1 is a basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neurogenin 1
Monoclonal
1.0 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q92886
NEUROG1
Full length human recombinant protein of NEUROG1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
OTI3F9
Unconjugated
NEUROG1
AKA; ATH4C; basic helix-loop-helix transcription factor; bHLHa6; cb260; chunp6899; class A basic helix-loop-helix protein 6; Helix-loop-helix protein mATH-4C; Math4C; Neurod3; NEUROG1; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; neurogenic differentiation factor 3; neurogenin; neurogenin 1; neurogenin related protein-1; neurogenin-1; Neurogenin-related protein 1; NGN; Ngn1; NGN-1; ngr1; transcription factor; Zath4c; zNgn1
Mouse
Affinity Chromatography
RUO
4762
-20°C, Avoid Freeze/Thaw Cycles