missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 189.00 - € 532.00
Specifications
| Antigen | Neurogenin-2 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658187
|
Novus Biologicals
NBP2-41195-0.025mg |
0.025 mg |
€ 189.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18106998
|
Novus Biologicals
NBP2-41195 |
0.1 mg |
€ 532.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurogenin-2 Polyclonal specifically detects Neurogenin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Neurogenin-2 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 63973 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human NGN2. The immunogen is located within the last 50 amino acids of NGN2. Amino Acid Squence: CAGSDMDYWQPPPPDKHRYA | |
| Primary | |
| NGN2 antibody is predicted to not cross-react with neurogenin 1. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, neurogenin 2, neurogenin-2, ngn-2, NGN2, Protein atonal homolog 4 | |
| NEUROG2 | |
| IgG | |
| Affinity Purified | |
| 30 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title