missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Neurogenin-3 Antibody (NGN3/1808) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26980FR
This item is not returnable.
View return policy
Description
Neurogenin-3 Monoclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Neurogenin-3 | |
| Monoclonal | |
| DyLight 680 | |
| Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 | |
| Recombinant fragment (around aa1-200) of human Neurogenin-3 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Stem Cells | |
| 50674 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| NGN3/1808 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction