missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 335.00
Specifications
| Antigen | Neurogenin-3 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:250-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610992
|
Novus Biologicals
NBP2-98714-100ul |
100 μg |
€ 335.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607012
|
Novus Biologicals
NBP2-98714-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurogenin-3 Polyclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Neurogenin-3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Stem Cells | |
| 0.2 um filtered solution in PBS | |
| 50674 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:250-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Neurogenin-3 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title