missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogenin-3 Antibody (OTI3B5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72956F
This item is not returnable.
View return policy
Description
Neurogenin-3 Monoclonal specifically detects Neurogenin-3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Neurogenin-3 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NEUROG3 | |
| Full-length protein expressed in 293T cell transfected with human Neurogenin-3 expression vector. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3B5 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 | |
| Mouse | |
| Affinity Purified | |
| Stem Cells | |
| 50674.0 | |
| Store at 4°C in the dark. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido