missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroligin 2/NLGN2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41299JF549
This item is not returnable.
View return policy
Description
Neuroligin 2/NLGN2 Polyclonal antibody specifically detects Neuroligin 2/NLGN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Neuroligin 2/NLGN2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human NLGN2. The immunogen is located within amino acids 630 - 680 of NLGN2. Amino Acid Squence: PPATLPPEPEPEPGPRAYD | |
| 0.1 mL | |
| Cognition and Behavior | |
| 57555 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction