missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroligin 4X/NLGN4X Antibody (S98-7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42198F
This item is not returnable.
View return policy
Description
Neuroligin 4X/NLGN4X Monoclonal specifically detects Neuroligin 4X/NLGN4X in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Neuroligin 4X/NLGN4X | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NLGN4X | |
| Fusion protein amino acids 782-945 (intracellular C-terminus) of mouse Neuroligin 4. ∼45% identity with Neuroligin-1, -2 and -3. | |
| 0.1 mL | |
| 57502 | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S98-7 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| ASPGX2, HLNX, HNLX, KIAA1260AUTSX2, MGC22376, neuroligin 4, X-linked, NLGN, NLGN4neuroligin 4, X-linked | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Approx 45% identity with Neuroligin-1, -2 and -3.Detects approx 25-130kDa. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction