missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuromedin BR/NMBR Antibody (466505), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4728CL1
This item is not returnable.
View return policy
Description
Neuromedin BR/NMBR Monoclonal antibody specifically detects Neuromedin BR/NMBR in Human samples. It is validated for Flow Cytometry,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Neuromedin BR/NMBR | |
| Monoclonal | |
| CoraFluor 1 | |
| neuromedin B receptor, neuromedin-B receptor, Neuromedin-B-preferring bombesin receptor, NMB-R | |
| NS0 mouse myeloma cell line transfected with human NMBR, Met1-Met390, Accession # AAB27330 | |
| 0.1 mL | |
| GPCR | |
| 4829 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 466505 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction