missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuron-specific beta-III Tubulin Biotinylated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems BAM1195
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Multi-Species samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| beta-III Tubulin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Beta3-tubulin, beta-4, betaIII Tubulin, beta-Tubulin III, Class III beta-tubulin, TUBB3, TUBB4 CFEOM3A, Tubulin Beta 3, tubulin beta-3 chain, Tubulin beta-4, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10381 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| TuJ-1 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| TUBB3 | |
| Rat brain-derived microtubules | |
| 100 μg | |
| Primary | |
| Detects mammalian and chicken neuron-specific beta-III tubulin but not other beta-tubulin isotypes. | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction