missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuronal Pentraxin R/NPTXR Antibody (12) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26911JF585
This item is not returnable.
View return policy
Description
Neuronal Pentraxin R/NPTXR Monoclonal antibody specifically detects Neuronal Pentraxin R/NPTXR in Human samples. It is validated for ELISA
Specifications
| Neuronal Pentraxin R/NPTXR | |
| Monoclonal | |
| Janelia Fluor 585 | |
| neuronal pentraxin receptor, NPR | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Neuronal Pentraxin R/NPTXR (RefSeq#: NP_055108.2; Ala24-Ala500). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 23467 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction