missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropeptide B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05567-20ul
This item is not returnable.
View return policy
Description
Neuropeptide B Polyclonal antibody specifically detects Neuropeptide B in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Neuropeptide B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| L7, neuropeptide B, PPL7, PPNPB, preproneuropeptide B, preproprotein L7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human Neuropeptide B (NP_683694.1). MARSATLAAAALALCLLLAPPGLAWYKPAAGHSSYSVGRAAGLLSGLRRSPYARRSQPYRGAEPPGGAGASPELQLHPRLRSLAVCVQDVAPNLQRCERLPDGRGTYQCKANVFLSLRAADCLAA | |
| 20 μg | |
| Neuroscience | |
| 256933 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction