missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropeptide FF R1/NPFFR1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS1903
This item is not returnable.
View return policy
Description
Neuropeptide FF R1/NPFFR1 Polyclonal antibody specifically detects Neuropeptide FF R1/NPFFR1 in Human, Porcine, Bovine, Equine, Primate, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Neuropeptide FF R1/NPFFR1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Pig, Bovine, Equine, Primate | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 19 μg/mL | |
| FLJ10751, G protein-coupled receptor 147, GPR147, neuropeptide FF 1, neuropeptide FF receptor 1, NPFF1G-protein coupled receptor 147, NPFF1R1, OT7T022, RFamide-related peptide receptor OT7T022 | |
| Synthetic 20 amino acid peptide from 2nd extracellular domain of human Neuropeptide FF R1/NPFFR1. | |
| 0.05 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 64106 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction