Learn More
Neuropilin-1 Antibody (130603) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB566JF669
Product Code. 30162133
  • € 812.00 / 0.50mL
Description
Neuropilin-1 Monoclonal antibody specifically detects Neuropilin-1 in Rat samples. It is validated for Flow Cytometry,Blockade of Receptor-ligand Interaction,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neuropilin-1
Monoclonal
Janelia Fluor 669
BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-1, Phe22-Asp854 (Lys811Arg, Pro812-Gly828 del), predicted, Accession # Q9QWJ9
RUO
Primary
Rat
Liquid
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
130603
50mM Sodium Borate
Mouse
0.5 mL
Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Phospho Specific, Virology Bacteria and Parasites
8829
Store at 4°C in the dark.
IgG2b