missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropilin-1 Antibody (130603), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB566PECY7
This item is not returnable.
View return policy
Description
Neuropilin-1 Monoclonal antibody specifically detects Neuropilin-1 in Rat samples. It is validated for Flow Cytometry
Specifications
| Neuropilin-1 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Flow Cytometry | |
| 130603 | |
| Flow Cytometry | |
| BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
| Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-1, Phe22-Asp854 (Lys811Arg, Pro812-Gly828 del), predicted, Accession # Q9QWJ9 | |
| 0.5 mL | |
| Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Phospho Specific, Virology Bacteria and Parasites | |
| 8829 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction