Learn More
Neuropilin-1 Antibody (761705) [FITC], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB5994F
Product Code. 18604824
  • € 547.00 / 0.10mL
Description
Neuropilin-1 Monoclonal antibody specifically detects Neuropilin-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neuropilin-1
Monoclonal
FITC
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry, CyTOF
761705
Flow Cytometry, CyTOF-ready
BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Mouse myeloma cell line NS0-derived recombinant mouse Neuropilin-1, Phe21-Pro856 , Accession # P97333
0.1 mL
Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Virology Bacteria and Parasites
8829
Store at 4°C in the dark.
IgG2a